Assay Details
| Gene Symbol: PTPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003432671.4 | XP_003432719.1 | 6 - 7 | 659 | 136 | |
| XM_005615980.3 | XP_005616037.1 | 5 - 6 | 787 | 136 | ||
| XM_005615982.3 | XP_005616039.1 | 6 - 7 | 659 | 136 | ||
| XM_533558.6 | XP_533558.2 | 5 - 6 | 787 | 136 | ||
More Information
Set Membership: |
|

PTPDC1
Genomic Map