Assay Details
| Gene Symbol: PWWP3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641623.3 | XP_005641680.1 | 6 | 2740 | 165 | |
| XM_005641624.3 | XP_005641681.1 | 6 | 2730 | 165 | ||
| XM_005641625.3 | XP_005641682.1 | 6 | 2669 | 165 | ||
| XM_005641626.3 | XP_005641683.1 | 7 | 2868 | 165 | ||
| XM_014111789.2 | XP_013967264.1 | 6 | 2698 | 165 | ||
| XM_022415609.1 | XP_022271317.1 | 7 | 2857 | 165 | ||
| XM_022415610.1 | XP_022271318.1 | 7 | 2809 | 165 | ||
| XM_022415611.1 | XP_022271319.1 | 6 | 2692 | 165 | ||
More Information
Set Membership: |
|

PWWP3B
Genomic Map