Assay Details
| Gene Symbol: CDC20B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022409329.1 | XP_022265037.1 | 9 - 10 | 1348 | 144 | |
| XM_022409330.1 | XP_022265038.1 | 9 - 10 | 1341 | 144 | ||
| XM_022409334.1 | XP_022265042.1 | 9 - 10 | 1346 | 144 | ||
| XM_022409340.1 | XP_022265048.1 | 9 - 10 | 3939 | 144 | ||
| XM_022409341.1 | XP_022265049.1 | 10 - 11 | 1664 | 144 | ||
| XM_022409342.1 | XP_022265050.1 | 10 - 11 | 3841 | 144 | ||
| XM_022409345.1 | XP_022265053.1 | 10 - 11 | 1920 | 144 | ||
| XM_022409346.1 | XP_022265054.1 | 10 - 11 | 3601 | 144 | ||
| XM_022409358.1 | XP_022265066.1 | 9 - 10 | 1349 | 144 | ||
| XM_535236.5 | XP_535236.3 | 9 - 10 | 1340 | 144 | ||
More Information
Set Membership: |
|

CDC20B
Genomic Map