Assay Details
| Gene Symbol: RBM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171748.1 | NP_001165219.1 | 2 - 3 | 210 | 174 | |
| GenBank mRNA | DN266656.1 | - | 2 - 3 | 210 | 174 | |
| DQ341269.1 | - | 2 - 3 | 210 | 174 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the NM_001171748.1 transcripts.
This assay is in the 5'UTR region of the NM_001171748.1 transcripts.
Set Membership: |
|

RBM12
Genomic Map