Assay Details
| Gene Symbol: SPAG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014121849.2 | XP_013977324.1 | 408 | 136 | ||
| XM_022406677.1 | XP_022262385.1 | 602 | 136 | |||
| XM_022406683.1 | XP_022262391.1 | 602 | 136 | |||
| XM_535163.5 | XP_535163.3 | 602 | 136 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SPAG6
Genomic Map