Assay Details
| Gene Symbol: NEGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420630.1 | XP_022276338.1 | 3 - 4 | 677 | 114 | |
| XM_850432.5 | XP_855525.2 | 3 - 4 | 677 | 114 | ||
More Information
Set Membership: |
|

NEGR1
Genomic Map