Assay Details
| Gene Symbol: ENPP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624032.3 | XP_005624089.1 | 270 | 54 | ||
| XM_022422654.1 | XP_022278362.1 | 270 | 54 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ENPP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624032.3 | XP_005624089.1 | 270 | 54 | ||
| XM_022422654.1 | XP_022278362.1 | 270 | 54 | |||
Set Membership: |
|