Assay Details
| Gene Symbol: HPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633731.3 | XP_005633788.1 | 15 - 16 | 2183 | 73 | |
| XM_005633732.3 | XP_005633789.1 | 15 - 16 | 2217 | 73 | ||
| XM_014122657.2 | XP_013978132.1 | 15 - 16 | 2183 | 73 | ||
| XM_022407913.1 | XP_022263621.1 | 10 - 11 | 1280 | 73 | ||
| XM_542523.5 | XP_542523.1 | 14 - 15 | 2097 | 73 | ||
More Information
Set Membership: |
|

HPS5
Genomic Map