Assay Details
| Gene Symbol: LTB4R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115797.2 | XP_013971272.1 | 2 | 2066 | 83 | |
| XM_014115798.2 | XP_013971273.1 | 2 | 2062 | 83 | ||
More Information
Set Membership: |
|

LTB4R
Genomic Map