Assay Details
| Gene Symbol: CRTC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622810.3 | XP_005622867.1 | 11 - 12 | 1531 | 91 | |
| XM_014115408.2 | XP_013970883.2 | 11 - 12 | 1461 | 91 | ||
| XM_014115409.2 | XP_013970884.2 | 11 - 12 | 1443 | 91 | ||
| XM_014115410.2 | XP_013970885.1 | 11 - 12 | 1510 | 91 | ||
More Information
Set Membership: |
|

CRTC2
Genomic Map