Assay Details
| Gene Symbol: PKIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433669.4 | XP_003433717.1 | 3 - 4 | 421 | 104 | |
| XM_005638060.3 | XP_005638117.2 | 2 - 3 | 191 | 104 | ||
| XM_005638062.2 | XP_005638119.1 | 3 - 4 | 344 | 104 | ||
| XM_022412474.1 | XP_022268182.1 | 2 - 3 | 287 | 104 | ||
| XM_848290.4 | XP_853383.2 | 2 - 3 | 281 | 104 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PKIA
Genomic Map