Assay Details
| Gene Symbol: PKM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433934.4 | XP_003433982.1 | 9 - 10 | 1460 | 72 | |
| XM_005638561.2 | XP_005638618.1 | 10 - 11 | 1640 | 72 | ||
| XM_014109434.2 | XP_013964909.2 | 11 - 12 | 1754 | 72 | ||
| XM_022412800.1 | XP_022268508.1 | 11 - 12 | 1749 | 72 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PKM
Genomic Map