Assay Details
| Gene Symbol: KIAA0319L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119247.2 | XP_013974722.1 | 16 - 17 | 2766 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA0319L| Gene Symbol: KIAA0319L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119247.2 | XP_013974722.1 | 16 - 17 | 2766 | 106 | |
Set Membership: |
|