Assay Details
| Gene Symbol: AXDND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622463.3 | XP_005622520.1 | 7 - 8 | 827 | 110 | |
| XM_014115152.2 | XP_013970627.1 | 7 - 8 | 904 | 110 | ||
| XM_022420987.1 | XP_022276695.1 | 7 - 8 | 904 | 110 | ||
| XM_022420988.1 | XP_022276696.1 | 7 - 8 | 904 | 110 | ||
| XM_022420989.1 | XP_022276697.1 | 6 - 7 | 697 | 110 | ||
| XM_022420990.1 | XP_022276698.1 | 7 - 8 | 904 | 110 | ||
| XM_022420991.1 | XP_022276699.1 | 5 - 6 | 502 | 110 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AXDND1
Genomic Map