Assay Details
| Gene Symbol: SLC16A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618894.3 | XP_005618951.1 | 2 - 3 | 358 | 173 | |
| XM_005618897.3 | XP_005618954.1 | 2 - 3 | 346 | 173 | ||
| XM_022417436.1 | XP_022273144.1 | 2 - 3 | 347 | 173 | ||
| XM_022417437.1 | XP_022273145.1 | 2 - 3 | 375 | 173 | ||
| XM_022417438.1 | XP_022273146.1 | 2 - 3 | 321 | 173 | ||
| XM_022417439.1 | XP_022273147.1 | 2 - 3 | 359 | 173 | ||
| XM_546110.5 | XP_546110.1 | 2 - 3 | 251 | 173 | ||
More Information
Set Membership: |
|

SLC16A9
Genomic Map