Assay Details
| Gene Symbol: RAB6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122665.2 | XP_013978140.1 | 5 - 6 | 841 | 123 | |
| XM_014122666.2 | XP_013978141.1 | 5 - 6 | 842 | 123 | ||
| XM_022407706.1 | XP_022263414.1 | 5 - 6 | 844 | 123 | ||
| XM_022407707.1 | XP_022263415.1 | 5 - 6 | 375 | 123 | ||
| XM_022407708.1 | XP_022263416.1 | 5 - 6 | 353 | 123 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAB6A
Genomic Map