Assay Details
| Gene Symbol: RGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640819.3 | XP_005640876.1 | 4 - 5 | 445 | 157 | |
| XM_014111294.2 | XP_013966769.1 | 4 - 5 | 546 | 157 | ||
| XM_545702.5 | XP_545702.4 | 4 - 5 | 540 | 157 | ||
| XM_853197.5 | XP_858290.1 | 4 - 5 | 441 | 157 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RGS1
Genomic Map