Assay Details
| Gene Symbol: ABCA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624281.3 | XP_005624338.1 | 2999 | 173 | ||
| XM_005624282.3 | XP_005624339.1 | 3179 | 173 | |||
| XM_005624285.3 | XP_005624342.1 | 3006 | 173 | |||
| XM_022422866.1 | XP_022278574.1 | 3090 | 173 | |||
| XM_022422867.1 | XP_022278575.1 | 2890 | 173 | |||
| XM_548020.6 | XP_548020.2 | 2999 | 173 | |||
More Information
Set Membership: |
|

ABCA8
Genomic Map