Assay Details
| Gene Symbol: SEPHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190410.1 | NP_001177339.1 | 273 | 163 | ||
| XM_014122200.2 | XP_013977675.1 | 408 | 163 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SEPHS1
Genomic Map