Assay Details
| Gene Symbol: ASB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623755.3 | XP_005623812.1 | 7 - 8 | 1471 | 68 | |
| XM_849378.5 | XP_854471.1 | 7 - 8 | 1469 | 68 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ASB2
Genomic Map