Assay Details
| Gene Symbol: PIP4K2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022424098.1 | XP_022279806.1 | 519 | 161 | ||
| XM_022424099.1 | XP_022279807.1 | 538 | 161 | |||
| XM_022424100.1 | XP_022279808.1 | 396 | 161 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PIP4K2C
Genomic Map