Assay Details
| Gene Symbol: LNX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628190.3 | XP_005628247.1 | 6 - 7 | 1660 | 61 | |
| XM_005628191.3 | XP_005628248.1 | 6 - 7 | 1675 | 61 | ||
| XM_022426991.1 | XP_022282699.1 | 7 - 8 | 3907 | 61 | ||
| XM_532373.6 | XP_532373.2 | 5 - 6 | 1565 | 61 | ||
| XM_844614.5 | XP_849707.1 | 6 - 7 | 2156 | 61 | ||
More Information
Set Membership: |
|

LNX1
Genomic Map