Assay Details
| Gene Symbol: RPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640599.3 | XP_005640656.1 | 3 - 4 | 339 | 144 | |
| XM_005640601.3 | XP_005640658.1 | 2 - 3 | 277 | 144 | ||
| XM_856221.5 | XP_861314.2 | 3 - 4 | 339 | 144 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RPE
Genomic Map