Assay Details
| Gene Symbol: GK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640255.2 | XP_003640303.1 | 7 - 8 | 795 | 168 | |
| XM_005641225.1 | XP_005641282.1 | 8 - 9 | 879 | 168 | ||
| XM_005641227.1 | XP_005641284.1 | 7 - 8 | 795 | 168 | ||
| XM_022415602.1 | XP_022271310.1 | 8 - 9 | 776 | 168 | ||
| XM_022415603.1 | XP_022271311.1 | 8 - 9 | 784 | 168 | ||
| XM_022415604.1 | XP_022271312.1 | 8 - 9 | 776 | 168 | ||
| XM_022415605.1 | XP_022271313.1 | 7 - 8 | 692 | 168 | ||
| XM_022415606.1 | XP_022271314.1 | 8 - 9 | 778 | 168 | ||
| XM_022415607.1 | XP_022271315.1 | 8 - 9 | 780 | 168 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GK
Genomic Map