Assay Details
| Gene Symbol: ZFP37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022424946.1 | XP_022280654.1 | 4 | 943 | 159 | |
| XM_022424947.1 | XP_022280655.1 | 4 | 885 | 159 | ||
More Information
Set Membership: |
|

ZFP37
Genomic Map