Assay Details
| Gene Symbol: DTNBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110569.2 | XP_013966044.1 | 8 | 590 | 144 | |
| XM_014110570.2 | XP_013966045.1 | 6 | 511 | 144 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DTNBP1
Genomic Map