Assay Details
Gene Symbol: ZNF654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638740.3 | XP_005638797.1 | 3608 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
