Assay Details
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616673.3 | XP_005616730.1 | 4 | 573 | 134 | |
XM_022403761.1 | XP_022259469.1 | 4 | 1004 | 134 | ||
XM_022403762.1 | XP_022259470.1 | 7 | 776 | 134 | ||
XM_022403765.1 | XP_022259473.1 | 5 | 712 | 134 | ||
XM_022403768.1 | XP_022259476.1 | 4 | 357 | 134 | ||
XM_022403773.1 | XP_022259481.1 | 4 | 1001 | 134 | ||
XM_022403782.1 | XP_022259490.1 | 5 | 709 | 134 | ||
XM_022403791.1 | XP_022259499.1 | 4 | 354 | 134 | ||
XM_541662.5 | XP_541662.2 | 4 | 570 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
