Assay Details
| Gene Symbol: RBM4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631426.3 | XP_005631483.1 | 2 | 419 | 162 | |
| XM_022405615.1 | XP_022261323.1 | 2 | 419 | 162 | ||
| XM_022405616.1 | XP_022261324.1 | 2 | 342 | 162 | ||
| XM_022405617.1 | XP_022261325.1 | 2 | 442 | 162 | ||
| XM_022405618.1 | XP_022261326.1 | 2 | 427 | 162 | ||
| XM_533216.6 | XP_533216.1 | 2 | 417 | 162 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RBM4B
Genomic Map