Assay Details
| Gene Symbol: ANXA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002961.1 | NP_001002961.1 | 10 - 11 | 842 | 125 | |
| GenBank mRNA | AY422991.1 | - | 10 - 11 | 842 | 125 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ANXA2
Genomic Map