Assay Details
| Gene Symbol: KRT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253744.1 | NP_001240673.1 | 1087 | 128 | ||
| GenBank mRNA | DN871766.1 | - | 614 | 128 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

KRT15
Genomic Map