Assay Details
| Gene Symbol: PODXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014121652.2 | XP_013977127.1 | 1702 | 68 | ||
| GenBank mRNA | DQ202369.1 | - | 1526 | 68 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PODXL2
Genomic Map