Assay Details
| Gene Symbol: HPCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617615.2 | XP_005617672.1 | 3 | 719 | 131 | |
| XM_022411818.1 | XP_022267526.1 | 3 | 605 | 131 | ||
| XM_849499.5 | XP_854592.3 | 3 | 590 | 131 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HPCA
Genomic Map