Assay Details
| Gene Symbol: KCNN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632711.3 | XP_005632768.2 | 7 - 8 | 1384 | 76 | |
| XM_005632712.3 | XP_005632769.2 | 7 - 8 | 1384 | 76 | ||
| XM_005632713.3 | XP_005632770.2 | 6 - 7 | 1374 | 76 | ||
| XM_005632714.3 | XP_005632771.1 | 8 - 9 | 1490 | 76 | ||
| XM_014122358.2 | XP_013977833.1 | 8 - 9 | 1388 | 76 | ||
| XM_022406880.1 | XP_022262588.1 | 6 - 7 | 1290 | 76 | ||
| XM_022406881.1 | XP_022262589.1 | 8 - 9 | 1434 | 76 | ||
| XM_541945.6 | XP_541945.5 | 6 - 7 | 1374 | 76 | ||
More Information
Set Membership: |
|

KCNN1
Genomic Map