Assay Details
| Gene Symbol: LZTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634860.3 | XP_005634917.1 | 4 | 1015 | 142 | |
| XM_014107229.2 | XP_013962704.1 | 4 | 1819 | 142 | ||
| XM_022409056.1 | XP_022264764.1 | 4 | 1051 | 142 | ||
| XM_022409057.1 | XP_022264765.1 | 4 | 975 | 142 | ||
| XM_022409058.1 | XP_022264766.1 | 4 | 953 | 142 | ||
| XM_022409059.1 | XP_022264767.1 | 4 | 1016 | 142 | ||
More Information
Set Membership: |
|

LZTS3
Genomic Map