Assay Details
| Gene Symbol: RUNX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413371.1 | XP_022269079.1 | 3 | 386 | 143 | |
| XM_022413372.1 | XP_022269080.1 | 3 | 296 | 143 | ||
| XM_022413373.1 | XP_022269081.1 | 3 | 396 | 143 | ||
| XM_022413374.1 | XP_022269082.1 | 3 | 402 | 143 | ||
More Information
Set Membership: |
|

RUNX1
Genomic Map