Assay Details
| Gene Symbol: GFI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012719.1 | NP_001012737.1 | 4 | 869 | 75 | |
| XM_022419500.1 | XP_022275208.1 | 4 | 930 | 75 | ||
| GenBank mRNA | AB178767.1 | - | 4 | 899 | 75 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GFI1
Genomic Map