Assay Details
| Gene Symbol: FIZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638798.4 | XP_003638846.1 | 3 | 662 | 149 | |
| XM_005616109.2 | XP_005616166.1 | 3 | 764 | 149 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FIZ1
Genomic Map