Assay Details
| Gene Symbol: MMP13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619857.3 | XP_005619914.1 | 7 - 8 | 1118 | 99 | |
| XM_022418390.1 | XP_022274098.1 | 9 - 10 | 1665 | 99 | ||
| XM_022418391.1 | XP_022274099.1 | 9 - 10 | 1431 | 99 | ||
| XM_536598.5 | XP_536598.3 | 8 - 9 | 1386 | 99 | ||
| GenBank mRNA | AF149850.1 | - | 2 - 3 | 235 | 99 | |
| AF201729.1 | - | 4 - 5 | 617 | 99 | ||
| LC028238.1 | - | 4 - 5 | 664 | 99 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MMP13
Genomic Map