Assay Details
| Gene Symbol: PAM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420135.1 | XP_022275843.1 | 2 - 3 | 248 | 75 | |
| XM_022420136.1 | XP_022275844.1 | 2 - 3 | 554 | 75 | ||
| GenBank mRNA | BQ235428.1 | - | 1 - 2 | 113 | 75 | |
More Information
Set Membership: |
|

PAM16
Genomic Map