Assay Details
| Gene Symbol: PAM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420135.1 | XP_022275843.1 | 4 - 5 | 444 | 82 | |
| XM_022420136.1 | XP_022275844.1 | 4 - 5 | 750 | 82 | ||
| GenBank mRNA | BQ235428.1 | - | 3 - 4 | 309 | 82 | |
More Information
Set Membership: |
|

PAM16
Genomic Map