Assay Details
| Gene Symbol: FUT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005378.1 | NP_001005378.1 | 1 | 935 | 82 | |
| XM_022406216.1 | XP_022261924.1 | 2 | 1429 | 82 | ||
| XM_022406217.1 | XP_022261925.1 | 2 | 1148 | 82 | ||
| GenBank mRNA | AJ831836.1 | - | 1 | 935 | 82 | |
| Gene Symbol: LOC100683739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003432795.4 | XP_003432843.1 | 1 | 1045 | 82 | |
More Information
Set Membership: |
|

FUT5
Genomic Map