Assay Details
| Gene Symbol: P2RY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193673.1 | NP_001180602.1 | 1 | 1066 | 76 | |
| GenBank mRNA | DN374749.1 | - | 1 | 443 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: P2RY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193673.1 | NP_001180602.1 | 1 | 1066 | 76 | |
| GenBank mRNA | DN374749.1 | - | 1 | 443 | 76 | |
Set Membership: |
|