Assay Details
| Gene Symbol: KCNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006645.2 | NP_001006646.1 | 1 | 1752 | 58 | |
| GenBank mRNA | U08596.1 | - | 1 | 58 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KCNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006645.2 | NP_001006646.1 | 1 | 1752 | 58 | |
| GenBank mRNA | U08596.1 | - | 1 | 58 | ||
Set Membership: |
|