Assay Details
| Gene Symbol: KRT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116588.2 | XP_013972063.1 | 1179 | 93 | ||
| XM_014116589.2 | XP_013972064.1 | 1179 | 93 | |||
| XM_014116590.2 | XP_013972065.1 | 1179 | 93 | |||
More Information
Set Membership: |
|

KRT10
Genomic Map