Assay Details
| Gene Symbol: CRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014278.2 | NP_001014300.2 | 2 | 1136 | 74 | |
| XM_005637934.3 | XP_005637991.1 | 3 | 920 | 74 | ||
More Information
Set Membership: |
|

CRH
Genomic Map