Assay Details
Gene Symbol: OR1E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014287.1 | NP_001014309.1 | 1 | 897 | 85 | |
XM_022422596.1 | XP_022278304.1 | 2 | 2258 | 85 | ||
XM_022422597.1 | XP_022278305.1 | 2 | 2235 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
