Assay Details
| Gene Symbol: SSTR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025620.1 | NP_001020791.1 | 1 | 767 | 135 | |
| XM_005625695.3 | XP_005625752.1 | 2 | 1410 | 135 | ||
More Information
Set Membership: |
|

SSTR3
Genomic Map