Assay Details
| Gene Symbol: ARSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048122.1 | NP_001041587.1 | 6 - 7 | 980 | 87 | |
| XM_005640996.3 | XP_005641053.1 | 7 - 8 | 1984 | 87 | ||
| XM_005640997.3 | XP_005641054.1 | 7 - 8 | 1598 | 87 | ||
| XM_005640998.3 | XP_005641055.1 | 7 - 8 | 1131 | 87 | ||
| XM_014111332.2 | XP_013966807.1 | 7 - 8 | 1166 | 87 | ||
More Information
Set Membership: |
|

ARSL
Genomic Map