Assay Details
| Gene Symbol: CASP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077236.1 | NP_001070704.1 | 4 - 5 | 595 | 84 | |
| GenBank mRNA | DQ228670.1 | - | 4 - 5 | 595 | 84 | |
| DQ228671.1 | - | 2 - 3 | 298 | 84 | ||
More Information
Set Membership: |
|

CASP12
Genomic Map